missing translation for 'onlineSavingsMsg'
Learn More

Spectrophotometry Holmium Oxide Solution UV and Visible Wavelength Standard 240 nm to 640 nm, Reagecon
BIOPHARMA

Used for validation of the wavelength scale of a spectrophotometer in both the UV and visible regions.

Brand:  Reagecon RSPEC00151


Product Code. 13502080

  • £122.00 / 100mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Spectrophotometry Reference Standard, Spectrophotometry Reference Material.
  • Provides peak wavelengths between 240 nm and 640 nm that includes a spectral scan at various wavelengths including 0.1, 0.2, 0.5, 1.0 and 2.0 nm slit widths.
  • Used for validation of the wavelength scale of a spectrophotometer in both the UV and Visible regions.
  • This spectrophotometry standard is prepared gravimetrically on a weight/weight basis.
  • Both solute and solvent are weighed on a balance calibrated by Reagecon engineers using OIML traceable weights.
  • Reagecon holds ISO 17025 accreditation for calibration of laboratory balances (INAB Ref:265C).
  • The resulting Balance Certificate of Calibration is issued in accordance with the requirements of ISO/IEC 17025.
  • The certified values of each standard are verified using a high performance spectrophotometer calibrated with NIST traceable, ISO Guide 34 Certified Standards.
Specifications

Specifications

Amber Glass Winchester Bottle
100 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • holmium oxide solution UV and visible wavelength standard 240nm tp 640nm

Signal Word
  • Danger

Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1

Hazard Statement
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : perchloric acid

SDS
Documents

Documents

Certificates
Special Offers

Special Offers