missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrum™ Spectra/Por™ 1-4 Standard RC Dialysis Membrane in Flat Sheets
Clear, flexible and sturdy Standard RC (regenerated cellulose) in single layer square sheets that can be used with a variety of dialysis equipment or can be cut into specific dimensions for attaching to support frames for customized diagnostic assays, equilibrium studies, desalting, protein purification, etc.
£272.11 - £451.00
Specifications
Color | Clear |
---|---|
Temperature Range (English) | 39°F to 250°F |
Temperature Range (Metric) | 4°C to 121°C |
Quantity | 25 Sheets |
For Use With (Application) | Ideal for molecular separation, protein binding studies, electrolyte removal, desalting |
Product Code | Brand | Membrane | Length (English) | MWCO | Width (English) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Membrane | Length (English) | MWCO | Width (English) | Price | Quantity & Availability | ||
10278854
|
Spectrum™
132686 |
Spectra/Por 2 | 7.8 in. | 12 to 14 kDa | 7.8 in. |
£404.00
Pack of 25 |
Estimated Shipment: 18-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
12757516
|
Spectrum™
132723 |
Spectra/Por 3 | 4.2 in. | 3.5 kDa | 4.2 in. |
£307.00
Pack of 25 |
Estimated Shipment: 03-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
12360170
|
Spectrum™
132677 |
Spectra/Por 1 | 9.4 in. | 6 to 8 kDa | 9.4 in. |
£451.00
Pack of 25 |
Estimated Shipment: 18-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
12370170
|
Spectrum™
132712 |
Spectra/Por 4 | 5.9 in. | 12 to 14 kDa | 5.9 in. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Description
- Flat-Sheets deal for use with dialysis equipment or cutting to size
- Available in 3.5 kD, 6-8 kD and 12-14 kD MWCO's
- Good chemical compatibility with dilute strong acids & bases, conc. weak acids and bases, many alcohols & some organics.
- Standard RC can tolerate pH 2 - 12 and temp 4-121°C.
- 25 sheets/pkg
Do not tie knots in tubing; use closures
Specifications
Clear | |
4°C to 121°C | |
Ideal for molecular separation, protein binding studies, electrolyte removal, desalting |
39°F to 250°F | |
25 Sheets |