Learn More
Spectrum™ Spectra/Por™ 6 Pre-wetted Standard RC Dialysis Tubing, 1-50 kD MWCO
Clear, flexible and sturdy; Standard Grade RC (regenerated cellulose) membrane conveniently pre-equilibrated in water, eliminating the need to soak to remove glycerin and hydrate the membrane
Brand:  Spectrum™ 132640
 View more versions of this product
Product Code. 11445869
  • £674.00 / Each
Explore more special offers
Description
Description
Clear, flexible and sturdy; Standard Grade RC (regenerated cellulose) membrane conveniently pre-equilibrated in water, eliminating the need to soak to remove glycerin and hydrate the membrane
This ready-to-use membrane is packaged wet in a preservative solution of 0.05% sodium azide. Simply rinse away the preservative solution with DI water, load the sample and dialyze.
  • Pre-wetted & ready-to-use, no glycerin removal required
  • 8 MWCO's ranging from 1 kD - 50 kD
  • Good chemical compatibility with dilute strong acids and bases, conc. weak acids & bases, many alcohols and some organics
  • Standard RC can tolerate pH 2 - 12 and temp 4-121°C.
  • Ideal for sample prep, desalting, buffer exchange, pH change, protein purification, etc.
  • Economic 10 meters/pkg
Do not tie knots in tubing; use closures
Specifications
Specifications
Spectra/Por 6 Dialysis Tubing
Regenerated Cellulose
1.14 in.
10 m
39.2°F to 249.8°F
Each
1 kDa
29 mm
33 ft.
Packaged wet in 0.05% sodium azide
4°C to 121°C
Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products