missing translation for 'onlineSavingsMsg'
Learn More

Spectrum™ Clear Bath

Product Code. 11455839
Click to view available options
Volume (Metric):
230 mL
Unit Size:
230mL
This item is not returnable. View return policy

Product Code. 11455839

Brand: Spectrum™ 105 540

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Keeps water baths, incubators, temperature controlled instruments, humidifiers, etc. clean and clear

  • Does not contain copper or other heavy metals

Specifications

Form Liquid
pH 6.5 to 8.3
Appearance Pale Yellow
Boiling Point 200°F
Certifications/Compliance US OSHA, CFR 1910.1200, WHMIS
Composition Benzalkonium Chloride, n-alkyl (50% C14, 40% C12, 10% C16) Dimethyl Benzyl Ammonium Chloride as acti
For Use With (Application) Used to keep any circulating, temperature-controlled instrument clean, instruments like circulating Water baths, incubators, temperature controlled instruments, humidifiers, automatic analyzer baths and Blood/gas analyzers
Volume (English) 8 oz.
Volume (Metric) 230 mL
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Clear Bath®
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P271-Use only outdoors or in a well-ventilated area.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P233-Keep container tightly closed.
  • P270-Do not eat, drink or smoke when using this product.
  • P264-Wash thoroughly after handling.

Contains State Restrictions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.