Learn More
Spectrum™ Spectra/Por™ 1 RC Dialysis Membrane Tubing 6000 to 8000 Dalton MWCO
Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
£356.00 - £777.00
Specifications
MWCO 6 to 8 kDa
Material Regenerated Cellulose
Temperature Range (English) 39.2°F to 249.8°F
Temperature Range (Metric) 4°C to 121°C
Quantity Each
View More Specs
Products 7
Product Code Brand Diameter (Metric) Length (Metric) Price Quantity & Availability  
Product Code Brand Diameter (Metric) Length (Metric) Price Quantity & Availability  
11425849
View accessory products View Documents
Spectrum™
132660
25.5 mm 30.5 m
£555.00
Each
11405849
View accessory products View Documents
Spectrum™
132650
14.6 mm 30.5 m
£356.00
Each
11415849
View accessory products View Documents
Spectrum™
132655
20.4 mm 30.5 m
£513.00
Each
11455849
View Documents
Spectrum™
132675
76 mm 15 m
£473.20
Each
11445849
View accessory products View Documents
Spectrum™
132670
64 mm 15 m
£777.00
Each
11495839
View Documents
Spectrum™
132645
6.4 mm 15.2 m
£502.00
Each
11435849
View accessory products View Documents
Spectrum™
132665
32 mm 30.5 m
£638.00
Each
Description
Description
Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
Supplied as flat-width tubing or flat sheets that can be folded to fit various funnel-type dialysis apparatus. Pure regenerated natural cellulose membranes with 1000 to 50,000 molecular weight cutoffs.
  • Transparent and flexible
  • Low molecular weight compounds in aqueous solutions diffuse through membrane
  • Higher molecular weights, such as proteins and bacteria, are retained by membrane
  • Glycerol preserves membrane and must be removed before use
  • Flat sheets and discs available for use with dialysis apparatus
Do not tie knots in tubing; use closures
Specifications
Specifications
6 to 8 kDa
39.2°F to 249.8°F
Each
Regenerated Cellulose
4°C to 121°C
Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Spectrum™ Spectra/Por™ 1 RC Dialysis Membrane Tubing 6000 to 8000 Dalton MWCO