Learn More
Spectrum™ Spectra/Por™ 2 RC Dialysis Membrane Tubing 12,000 to 14,000 Dalton MWCO
Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
Brand:  Spectrum™ 132676
 View more versions of this product
Product Code. 11465849
  • £505.00 / Each
Explore more special offers
Description
Description
Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
Supplied as flat-width tubing or flat sheets that can be folded to fit various funnel-type dialysis apparatus. Pure regenerated natural cellulose membranes with 1000 to 50,000 molecular weight cutoffs.
  • Transparent and flexible
  • Low molecular weight compounds in aqueous solutions diffuse through membrane
  • Higher molecular weights, such as proteins and bacteria, are retained by membrane
  • Glycerol preserves membrane and must be removed before use
  • Flat sheets and discs available for use with dialysis apparatus
Do not tie knots in tubing; use closures
Specifications
Specifications
Spectra/Por 2 Dialysis Tubing
Regenerated Cellulose
0.25 in.
15 m
39.2°F to 249.8°F
Each
12 to 14 kDa
6.4 mm
49.2 ft.
Contain glycerin as a humectant
4°C to 121°C
Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products