missing translation for 'onlineSavingsMsg'
Learn More

Spectrum™ Spectra/Por™ 3 RC Dialysis Membrane Tubing 3500 Dalton MWCO

Ideal for molecular separation, protein binding studies, electrolyte removal, desalting

£616.00 - £663.00

Specifications

Product Type Spectra/Por 3 Dialysis Tubing
MWCO 3.5 kDa
Material Regenerated Cellulose
Length (English) 49.2 ft.
Length (Metric) 15 m
View More Specs

Products 3
Product Code Brand Diameter (Metric) Price Quantity & Availability  
Product Code Brand Diameter (Metric) Price Quantity & Availability  
10142634
View accessory products View Documents
Spectrum™
132724
29 mm
£633.00
Each
Please to purchase this item. Need a web account? Register with us today!
11425859
View accessory products View Documents
Spectrum™
132720
11.5 mm
£616.00
Each
Please to purchase this item. Need a web account? Register with us today!
11435859
View accessory products View Documents
Spectrum™
132725
34 mm
£663.00
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Ideal for molecular separation, protein binding studies, electrolyte removal, desalting

Supplied as flat-width tubing or flat sheets that can be folded to fit various funnel-type dialysis apparatus. Pure regenerated natural cellulose membranes with 1000 to 50,000 molecular weight cutoffs.

  • Transparent and flexible
  • Low molecular weight compounds in aqueous solutions diffuse through membrane
  • Higher molecular weights, such as proteins and bacteria, are retained by membrane
  • Glycerol preserves membrane and must be removed before use
  • Flat sheets and discs available for use with dialysis apparatus

Do not tie knots in tubing; use closures

Specifications

Specifications

Spectra/Por 3 Dialysis Tubing
Regenerated Cellulose
15 m
39.2°F to 249.8°F
Each
3.5 kDa
49.2 ft.
Contain glycerin as a humectant
4°C to 121°C
Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Spectrum™ Spectra/Por™ 3 RC Dialysis Membrane Tubing 3500 Dalton MWCO

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.