Learn More
Spectrum™ Spectra/Por™ 7 Membrane Tubing (Sulfur and Heavy Metal Free):1 1000 to 50,000 Dalton MWCO
Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
Brand:  Spectrum™ 132109
 View more versions of this product
Product Code. 11445909
  • £591.00 / Each
Explore more special offers
Description
Description
Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
Supplied as flat-width tubing or flat sheets that can be folded to fit various funnel-type dialysis apparatus. Pure regenerated natural cellulose membranes with 1000 to 50,000 molecular weight cutoffs.
  • Transparent and flexible
  • Low molecular weight compounds in aqueous solutions diffuse through membrane
  • Higher molecular weights, such as proteins and bacteria, are retained by membrane
  • Glycerol preserves membrane and must be removed before use
  • Flat sheets and discs available for use with dialysis apparatus
Do not tie knots in tubing; use closures
Specifications
Specifications
Spectra/Por 7 Membrane Tubing
Regenerated Cellulose
1.14 in.
5 m
Packaged wet in 0.05% sodium azide
4°C to 121°C
Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
2 kDa
29 mm
16.4 ft.
Transparent
39.2°F to 249.8°F
Each
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products