Learn More
Spectrum™ Spectra/Por™ 7 Membrane Tubing (Sulfur and Heavy Metal Free):1 1000 to 50,000 Dalton MWCO
Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
Brand: Spectrum™ 132125
Code : LB
Additional Details : Weight : 0.01000kg
Description
Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
Supplied as flat-width tubing or flat sheets that can be folded to fit various funnel-type dialysis apparatus. Pure regenerated natural cellulose membranes with 1000 to 50,000 molecular weight cutoffs.
- Transparent and flexible
- Low molecular weight compounds in aqueous solutions diffuse through membrane
- Higher molecular weights, such as proteins and bacteria, are retained by membrane
- Glycerol preserves membrane and must be removed before use
- Flat sheets and discs available for use with dialysis apparatus
Do not tie knots in tubing; use closures
Specifications
Spectra/Por 7 Membrane Tubing | |
Regenerated Cellulose | |
0.29 in. | |
5 m | |
Packaged wet in 0.05% sodium azide | |
4°C to 121°C | |
Ideal for molecular separation, protein binding studies, electrolyte removal, desalting |
25 kDa | |
7.5 mm | |
16.4 ft. | |
Transparent | |
39.2°F to 249.8°F | |
Each |