missing translation for 'onlineSavingsMsg'
Learn More

Spectrum™ Spectra/Por™ Biotech Cellulose Ester (CE) Dialysis Membrane Tubing

Selected MWCOs from 100 to 100,000 Daltons

Brand:  Spectrum™ 131096

 View more versions of this product

Product Code. 11405939

  • £1060.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Selected MWCOs from 100 to 100,000 Daltons

  • CE tubing membranes are ideal for desalting, purification, and molecular separation
  • Supplied in 10m (33 ft.) lengths in two rolls of 5m each, in 0.1% sodium azide solution

Order Info

Note:Do not tie—must be used with Spectra/Por nylon closures (08-671-52, -53, -55). Strong polar solvents such as acetone, MEK, or dioxane will irreparably damage membranes. Contact Spectrum Technical Service, 1-800-634-3300, for more information.

Specifications

Specifications

Spectra/Por Dialysis Membrane Tubing
Cellulose Ester
0.78 in.
10 m
39.2°F to 98.6°F
Each
1 kDa
20 mm
33 ft.
Supplied in 0.1% sodium azide solution
4°C to 37°C
Ideal for desalting, purification, and molecular separation
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers