missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrum™ Spectra/Por™ Biotech Cellulose Ester (CE) Dialysis Membrane Tubing
Selected MWCOs from 100 to 100,000 Daltons
Brand: Spectrum™ 131084
This item is not returnable.
View return policy
Description
Selected MWCOs from 100 to 100,000 Daltons
- CE tubing membranes are ideal for desalting, purification, and molecular separation
- Supplied in 10m (33 ft.) lengths in two rolls of 5m each, in 0.1% sodium azide solution
Order Info
Note:Do not tie—must be used with Spectra/Por nylon closures (08-671-52, -53, -55). Strong polar solvents such as acetone, MEK, or dioxane will irreparably damage membranes. Contact Spectrum Technical Service, 1-800-634-3300, for more information.
Specifications
Spectra/Por Dialysis Membrane Tubing | |
Cellulose Ester | |
0.25 in. | |
10 m | |
39.2°F to 98.6°F | |
Each |
1 kDa | |
6.4 mm | |
33 ft. | |
Supplied in 0.1% sodium azide solution | |
4°C to 37°C | |
Ideal for desalting, purification, and molecular separation |