missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrum™ Standard RC Ready-to-use Spectra/Por™ Dialysis Sacks
For large sample volumes
£281.60 - £352.00
Specifications
Product Type | Dialysis Sacks |
---|---|
Material | Regenerated Cellulose |
Length (English) | 23.6 in. |
Length (Metric) | 600 mm |
Capacity (Metric) | 50 to 400 mL |
Product Code | Brand | Diameter (English) | Membrane | MWCO | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (English) | Membrane | MWCO | Price | Quantity & Availability | |||
15330762
|
Spectrum™
132681 |
1.14 in. | Spectra/Por 2 | 12 to 14 kDa |
£312.00
Pack of 10 |
Estimated Shipment: 02-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15390772
|
Spectrum™
132726 |
1.33 in. | Spectra/Por 3 | 3.5 kDa |
£352.00
Pack of 10 |
Estimated Shipment: 14-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15310772
|
Spectrum™
132 707 |
1.14 in. | Spectra/Por 4 | 12 to 14 kDa |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
- Material: Regenerated Cellulose
- Length: 60cm (23.62 in.)
- Sealed at one end, open at the other
- Funnel attached to top for easy sample filling
- Supplied in sodium azide 0.05% (must be rinsed prior to use)
Specifications
Dialysis Sacks | |
23.6 in. | |
50 to 400 mL | |
For large sample volumes, funnel attached to top for easy sample filling |
Regenerated Cellulose | |
600 mm | |
0.05% sodium azide preservant |