Learn More
Cole-Parmer™ Specwell™ M820-S Microscope/Telescope with Built-in Reticle
Ideal for macroscopic quality control and inspection procedures. This scope is constructed of durable aluminum and plastic and features a straight line reticle.
Brand:  Cole-Parmer™ 03097-00
Product Code. 11759293
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Scope are constructed of durable aluminum and plastic
  • Lenses come with covers to protect against scratches
  • Eyepiece has a soft edge for comfort and safety
  • Includes a neck strap, a micro lens stand, hard carrying case and carrying strap
  • Ideal for macroscopic quality control and inspection procedures, this model includes a straight line reticle
  • For microscopic viewing, screw the micro lens and stand onto the monocular
  • For distant telescopic viewing, unscrew the micro lens and stand and use the monocular as a telescope (these scopes are not intended for close range telescopic viewing)
  • Focus is adjustable in both microscope and telescope modes
Specifications
Specifications
Microscope/Telescope with Built-in Reticle
2.7 mm
7.08 in.
Neck strap, a micro lens stand, hard carrying case and carrying strap
Specwell™ M820-S
1.06 in.
9 mm
18 cm
8x/25x
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cole-Parmer™ Specwell™ M820-S Microscope/Telescope with Built-in Reticle > Microscope/Telescope with Built-in Reticle