Learn More
Cytiva SpeedBeads™ Magnetic Neutravidin Coated Particles, 1 um, 1% solids, 0.05% Na azide
Capture of Biotinylated Probes. Their “cauliflower-like” structure gives Neutravidin beads high binding capacity. Neutravidin SpeedBeads combine fast reaction kinetics and low, non-specific binding for increased throughput and precision in immunoassay and molecular biology applications.
Brand: Cytiva 78152104010350
4347.20 GBP valid until 2024-12-31
Use promo code "21948" to get your promotional price.
Code : GE
Additional Details : Weight : 0.31750kg
Description
- Neutravidin beads exhibit high affinity when binding to biotinylated ligands such as proteins, nucleic acids and peptides. They respond quickly to a magnetic field to shorten assay times, improve precision and move faster through viscous solutions
- Available with a 3500-4500 pmol/mg biotin-binding range
Specifications
1% solids (10mg/mL) | |
Immunoassays, cDNA Library Construction, Capture of Biotinylated Targets | |
100 mL |
2°C to 8°C | |
0.05% Na azide | |
1 μm |