Learn More
3M™ Speedglas™ Head Protection 9100 FX
3M™ Speedglas™ Head Protection 9100 FX for use with Speedglas™ Welding Helmets 9100 FX and 9100 FX Air.
Brand: 3M™ 7100167240
72.65 GBP valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Additional Details : Weight : 1.00000kg
Description
- Made in flame-retardant textiles for protection against sparks and flames
- Extended head coverage, for use with the complete helmet assembly to help protect the user from welding spark and spatter on the top portion of the head
- As the owner of a Speedglas™ welding helmet, you have a variety of accessories available to help maximize your protection and comfort
Specifications
Black | |
Speedglas Welding Helmets 9100 FX and 9100 FX Air | |
Head Protection | |
Large |