missing translation for 'onlineSavingsMsg'
Learn More

3M™ Speedglas™ Hood

Product Code. 15780018
Click to view available options
:
Color: Black
This item is not returnable. View return policy

Product Code. 15780018

Brand: 3M™ 7100003083

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

3M™ Speedglas™ Hood offers extended head and neck protection for Speedglas™ Welding Helmets.

  • For use with the complete helmet assembly to help protect the user from welding spark, spatter, and radiation on the ears, neck and on the top portion of the head
  • Made in flame-retardant textiles for protection against molten metal, sparks and flames
  • Provides extended head and neck protection
TRUSTED_SUSTAINABILITY

Specifications

Color Black
For Use With (Equipment) Speedglas Welding Masks
Product Type Hood
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.