missing translation for 'onlineSavingsMsg'
Learn More

3M™ Speedglas™ Neck Protection

Product Code. p-7160340
Click to view available options
For Use With (Equipment):
Speedglas Welding Helmet Series 100, 9000, SL and 9100 MP
Speedglas Welding Helmet Series 9100
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15799757

Brand: 3M™ 7000044371

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

3M™ Speedglas™ Neck Protection offers extended protection for Speedglas™ Welding Helmet.

  • Made in flame-retardant textiles for protection against sparks and flames
  • For use with the complete helmet assembly to help protect the user from welding spark, spatter, and radiation on the ears and neck
TRUSTED_SUSTAINABILITY

Specifications

Color Black
For Use With (Equipment) Speedglas Welding Helmet Series 100, 9000, SL and 9100 MP
Product Type Neck Protection
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.