Learn More
3M™ Speedglas™ Outer Protection Plate 9000
3M™ Speedglas™ Outer Protection Plate 9000 for use with 3M™ Speedglas™ Welding Helmet Series 9000.
Brand: 3M™ 7000000206
47.30 GBP valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Code : NEW
Additional Details : Weight : 2.40000kg
Description
- Replacement spare part to the original protection plate if any pits or scratches occur
- For maximum safety and comfort, make it a habit to check your equipment regularly
- Regularly clean with a soft cloth and replace if any pits or scratches occur
Specifications
EN 166, EN166:BT | |
Clear | |
Heat Resistant |
Speedglas Welding Helmet Series 9000 | |
Helmet Outside Protection Plate |