Learn More
3M™ Speedglas™ Outside Protection Plate
3M™ Speedglas™ Outside Protection Plates are designed for use with 3M™ Speedglas™ Welding Helmets and Filters and protect the welding filter from sparks and spatter.
Brand: 3M™ 7100092252
220.00 GBP valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Additional Details : Weight : 2.00000kg
Description
- 3M™ Speedglas™ Outside Protection Plates are a replacement part designed to be used when the original has been damaged or worn out.
- Help protect your welding filter from sparks and spatter by ensuring that your Outside Protection Plate is functioning properly.
Specifications
ANSI Z89.1-2010 | |
9002NC Welding Helmets, 9100MP Welding Helmets, G5-02 Welding Helmets | |
Standard |
Clear | |
Outside Protection Plate |