Learn More
3M™ Speedglas™ Protective Visor Plate 9000 Flexview
3M™ Speedglas™ Protective Visor Plate 9000 Flexview is a replacement part for when visibility is reduced from pits and scratches.
Brand: 3M™ 7100080226
72.63 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Replacement spare part to the original visor plate if any pits or scratches occur
- For maximum safety and comfort, make it a habit to check your equipment regularly
- Replace worn out or damaged parts
- Regularly clean with a soft cloth
Specifications
Clear | |
Protective Visor Plate |