Learn More
Thermo Scientific™ SpeedVac™ SRF110 Refrigerated Centrifugal Vacuum Concentrator and Rotors
Process heat sensitive biological samples with confidence using SRF110. This Refrigerated SpeedVac system uses state-of-the-art centrifugation, vacuum, and heat technologies for removing solvents and concentrating heat sensitive biological samples while maintaining sample integrity.
£244.10 - £19409.86
Product Code | Brand | Description | Certifications | Material | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Certifications | Material | Price | Quantity & Availability | ||||
17364641
|
Thermo Scientific™
RF108 |
12-13 mm Rotor, Holds: 40 x 1.5, 2.0 mL; 16 x 3 mL | None | Aluminum |
£1059.49
Each |
In Stock
Log in to see stock. |
||||
17374641
|
Thermo Scientific™
RF132 |
Hexagonal Microcentrifuge Rotor; Holds: 132 x 1.5 mL microcentrifuge tube; 132 x 2.0 mL microcentrifuge tube | None | Aluminum |
£244.10
Each |
In Stock
Log in to see stock. |
||||
17384641
|
Thermo Scientific™
RF32 |
28mm rotor, Holds: 32 x 1.5 mL; 32 x 2 mL | None | Aluminum |
£974.73
Each |
Estimated Shipment: 29-07-2025
Log in to see stock. |
||||
17394641
|
Thermo Scientific™
RF52 |
12-17 mm rotor; Holds: 52 x 2 mL; 52 x 5 mL; 36 x 5-6mL | None | Aluminum |
£1110.34
Each |
In Stock
Log in to see stock. |
||||
17304651
|
Thermo Scientific™
RFMT4 |
MicroPlate Rotor; Holds: 4 x standard microplate; 2 deep well plates | None | Aluminum |
£838.27
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17314651
|
Thermo Scientific™
SRF110-230 |
Refrigerated, medium capacity, modular centrifugal vacuum concentrator, 230 V, 50 Hz, 8 Amps | CE, UKCA, KC, RCM | Powder coated steel |
£9493.03
Each |
In Stock
Log in to see stock. |
||||
17324651
|
Thermo Scientific™
SRF110P1-230 |
Medium capacity refrigerated centrifugal vacuum concentrator kit with oil pump, refrigerated vapor trap, rotor, accessories, 230 V, 50 Hz, 8 Amps | CE, UKCA, KC, RCM | Powder coated steel |
£17884.20
Each |
Estimated Shipment: 22-08-2025
Log in to see stock. |
||||
17334651
|
Thermo Scientific™
SRF110P2-230 |
Medium capacity refrigerated centrifugal vacuum concentrator kit with universal vacuum system, rotor, accessories, 230 V, 50 Hz, 8 Amps | CE, UKCA, KC, RCM | Powder coated steel |
£19409.86
Each |
Estimated Shipment: 22-08-2025
Log in to see stock. |
||||
Description
SpeedVac™ SRF110 modular refrigerated vacuum concentrator provides flexibility to your lab. It offers unique programming capabilities to maintain chamber temperature pre and post sample run to prevent thermal degradation of heat sensitive samples such as proteins, enzymes, and other biomolecules. They are ideal for evaporation of aqueous based solvents, buffers, and HPLC fractions. For a convenient, one model shopping experience select from SRF110P1 (modular configuration) and SRF110P2 (Semi integrated configuration) kit options. Each kit is designed to include components required for convenient SpeedVac setup.
SRF110 modular refrigerated vacuum concentrator provides flexibility to any benchtop space. SRF110 can be configured depending on your applications need. Select from wide range of suitable Thermo Scientific components such as pumps, Vapor Traps, and additional accessories to configure your custom vacuum concentration setup. For a convenient, one model shopping experience select from SRF110P1 (modular configuration) and SRF110P2 (Semi integrated configuration) kit options. Each kit is designed to include components required for convenient SpeedVac setup.
- Ideal for evaporation of non-aggressive, aqueous based solvents, buffers, HPLC fractions, etc.
- 3 Preset program with option to store up to 9 programs
- Temperature range -4°C up to 100°C (1°C increment)
- Self aligning and quick release rotor mechanism
- Hexagonal rotor (RF132) included with purchase of SRF110
- Refer to list of rotors available for purchase and to be used with SRF110 model
- Electricals- 115V AC (60Hz, 16Amps) and 230V AC (50Hz, 8Amps)
- Certifications- UL Certified, CE, UKCA, KC, RCM Marked.
- F Gas Compliance- Yes (EPA-SNAP and EU)
- SRF110P1 kit includes Refrigerated SpeedVac (SRF110), Hexagonal rotor (RF132), an Oil pump (VLP120-115), a vapor Trap (RVT450-115), Universal Tubing (UTP-TYG), 4L capacity Glass Condensation Flask (GCF400), Glass Cap (FC400), and 1L Bottle of heat transfer fluid Cryocool™ (SCC1)
- SRF110P2 kit includes SRF110P2 Kit includes Refrigerated SpeedVac (SRF110), Hexagonal rotor (RF132), Universal Vacuum System (UVS450), Universal Tubing (UTP-TYG), 4L capacity Glass Condensation Flask (GCF400), Glass Cap (FC400), and 1L Bottle of heat transfer fluid Cryocool™ (SCC1)