missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spermine tetrahydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
100 mg
Description
Polyamine which produces a variety of modulatory effects on the NMDA receptor channel, acting through a specific site on the complex which can cause both agonist and antagonist effects.

Specifications
Specifications
CAS | 306-67-2 |
Molecular Formula | C10H30Cl4N4 |
MDL Number | MFCD00012914 |
Quantity | 100 mg |
Synonym | spermine tetrahydrochloride, n,n'-bis 3-aminopropyl-1,4-butanediamine tetrahydrochloride, gerontine tetrahydrochloride, neuridine tetrahydrochloride, n1,n1'-butane-1,4-diyl bis propane-1,3-diamine tetrahydrochloride, musculamine tetrahydrochloride, geontine tetrahydrochloride, spermine hcl, 1,4-butanediamine, n,n'-bis 3-aminopropyl-, tetrahydrochloride, spermine, tetrahydrochloride |
InChI Key | XLDKUDAXZWHPFH-UHFFFAOYSA-N |
SMILES | [H+].[H+].[H+].[H+].[Cl-].[Cl-].[Cl-].[Cl-].NCCCNCCCCNCCCN |
IUPAC Name | tetrahydrogen (3-aminopropyl)({4-[(3-aminopropyl)amino]butyl})amine tetrachloride |
Molecular Weight (g/mol) | 348.18 |
PubChem CID | 9384 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction