Learn More
Cole-Parmer™ Disposable Closed XRF X-Cell Sample Cups
Use these Disposable Closed XRF X-Cell Sample Cups for fast, dependable, and reproducible XRF analysis of liquid samples.
£115.00 - £184.00
Specifications
Type | Sample Cup |
---|---|
Material | Polyethylene |
Quantity | 100 Pk. |
Requires: Window film
Product Code | Brand | Diameter (English) | Diameter (Metric) | Top Style | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (English) | Diameter (Metric) | Top Style | Price | Quantity & Availability | |||
15390552
|
Cole-Parmer™
3527 |
1.57 in. | 40 mm | Closed |
£145.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15300562
|
Cole-Parmer™
3529 |
1.2 in. | 31 mm | Closed |
£184.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15320562
|
Cole-Parmer™
3565 |
1.7 in. | 43 mm | Closed |
£172.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12884611
|
Cole-Parmer™
3571 |
1.2 in. | 31 mm | Double Open Ended, Collar |
£115.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15330562
|
Cole-Parmer™
3577 |
1.2 in. | 31 mm (micro) | Open, Collar |
£149.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- One opening for fast liquid sample delivery.
- Compatible with major suppliers of XRF spectrometers.
- Available in 31 mm, 40 mm, and 43 mm outer diameters.
- Contaminant-free polyethylene construction.
- Collars available to attach window film to cup for added security.
Cole-Parmer™ X-Cells are used throughout the world for fast, dependable, reproducible running of liquid samples in XRF spectrometers.
- They are designed to fit into the major suppliers of XRF spectrophotometers, such as PANalytical, Rigaku, Bruker, and Thermo Instruments.
- The durable polyethylene construction is generally resistant to acids, bases, and most organic chemicals except halogenated hydrocarbons.
- The patented closed X-Cell design features a snap-open seal for venting volatile samples and a reservoir cup for catching any liquids that expands through the vent.
- This assures the analyst of a smooth, fat window for improved precision, yet guards against spillage in inverted-optic spectrometers.
- A roughed surface within the reservoir cup facilitates labeling.
- Each closed X-Cell consists of a cup and snap-on ring.
- For routine liquid samples, the cup is filled with liquid, a piece of film is spready over the top, then the snap-ring is pushed down over the cup to form a fat drum-like window.
- The cell is turned over and, if desired, the snap-post on the rear can be pushed to break the seal and to permit venting of trapped bubbles and expand liquid while the sample is run.
- The filled X-Cell is then positioned in the spectrometer mask.
- Use the bubble-free insert with the 3527 (04576-05) 40 mm XRF liquid cell to prevent trapped air from forming a bubble under the window film when the cell is upright.
- The insert is pushed into the cell before the sample is added and forms an inverted-cone shelf under which air bubbles can be trapped once the cell is filled and sealed.
Order Info
Special application versions including the 31 mm micro X-Cell—with a 6.3 mm centered window for small, hazardous, or air-sensitive samples—and the 43 mm cell for Horiba™ sulfur analyzers also available. Supplied in bags of 100 cups.
Specifications
Sample Cup | |
Polyethylene | |
100 Pk. |