Learn More
SPEX CertiPrep™ OdorEroder
Effectively neutralizes a wide range of chemical odors in the lab
£41.02 - £41.50
Specifications
For Use With (Application) Effectively neutralizes a wide range of chemical odors in the lab
View More Specs
Products 2
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
15286106
View Documents
SPEX CertiPrep™
ODER-100G
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15296106
View Documents
SPEX CertiPrep™
ODER-250G
250 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • Non-toxic, environmentally safe purple beads trap odors, and convert them into harmless compounds that remain trapped in the beads
  • As odors get trapped, the beads start turning brown
Specifications
Specifications
Effectively neutralizes a wide range of chemical odors in the lab
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
SPEX CertiPrep™ OdorEroder