missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPEX CertiPrep™ Vial Holder
Thick, sturdy foam block that holds 24 standard 2ml tubes in the MiniG homogeniser from SPEX SamplePrep.
Brand: SPEX CertiPrep™ 1680
This item is not returnable.
View return policy
Description
- Rugged foam block to hold sample tubes securely in 1690 adjustable clamp for the 1600 MiniG.
- Process 24 samples at one time.
Specifications
Tubes (2 mL) | |
Foam Block | |
1690 adjustable clamp for the 1600 MiniG | |
24 |