missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Grinding Vial Set, Mid-Size Vial, PC Cylinder and End Plugs, PC Encapsulated Steel Impactor

This Mid-Size Grinding Vial has a sample capacity of up to 25 mL (1 to 40 g). Stainless steel end plugs and impactor and polycarbonate center cylinder.

Brand:  Cole-Parmer™ 6885-SSP

Product Code. 15380482

  • £860.00 / 1 set
Estimated Shipment: 15-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Grinds one sample at a time in the Large Freezer/Mill™ when used with the 6886 Mid-Size Grinding Vial Adapter.
  • Sample grinding capacity 3 g – 25 g.
  • Ideal for grinding soft plant and animal tissues and eliminates metal contamination.
  • Withstands liquid nitrogen temperatures.
  • Closed grinding vial prevents sample cross-contamination, retains volatiles, and controls hazardous or critical samples.
  • Transparent plastic center cylinder allows visual checking of the grinding progress.
  • Polycarbonate center cylinders are disposable.
  • Polycarbonate end plugs, impactor, and center cylinders may be cleaned with hot water and detergent or RNase/DNase-free solution.
  • Compatible with 6804 Large Extractor/Vial Opener when used in conjunction with 6884 Mid-Size Grinding Vial Adapter.
Specifications

Specifications

0.85 oz.
For Grinding Soft Tissues Without Metal Contamination
Polycarbonate
25 mL (0.85 oz.)
6870 Freezer/Mill
25 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers