Learn More
SPEX™ SamplePrep Large-Capacity Freezer/Mill™ Vial Divider for Grinding Vial Set 6751
For use with SPEX SamplePrep™ Large-Capacity Freezer/Mill Cryogenic Grinder
Brand: SPEX™ SamplePrep 6807
Code : NEW
Additional Details : Weight : 1.00000kg
Description
- Makes one complete vial plus three spare center sections
- Sample capacity: 0.5 to 4.0mL
- Includes stainless-steel impactor, two end plugs
Specifications
Multi Vial Adapter | |
Ideal for high throughput applications | |
Spex™ SamplePrep™ large capacity freezer/mill | |
Microvial set |