Learn More
Cole-Parmer™ XRF Prep-Aid™ Grinding Aids and Sample Binders
Prepare samples for reproducible and contamination-free results with XRF Prep-Aid™ Grinding Aids and Sample Binders.
Brand:  Cole-Parmer™ 3646-450
Code : NEW
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 15380552
  • £93.95 / 450g
In Stock
to see stock.
Description
Description
Ideal for samples which crumble or ablate after pressing.
  • Select binders from powder, tablet, or liquid form depending on your application.
  • High-quality sample preparation is the key to achieving consistent and reliable analytical results.
  • Powders are ideal for samples that crumble or ablate after pressing.
  • They blend quickly and completely with samples to form XRF (x-ray fluorescence) sample disks.
  • Tablets are an efficient way to add binder to XRF sample disks.
  • Liquid easily mixes with the sample.
Specifications
Specifications
Paraffin Binder
Samples Which Crumble or Ablate After Pressing
≤20 μm
450 g