missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sphingomyelin, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Description
Sphingomyelin is a eukaryotic sphingolipid and one of the major constituents of cell membranes and particularly abundant in the myelin sheath that surrounds neuronal axons. Sphingomyelin plays an important role in cell processes, the regulation of inflammatory responses, and signal transduction. Sphingomyelin metabolism is associated with various central nervous system diseases and Niemann-Pick disease.
Specifications
Specifications
| Chemical Name or Material | Sphingomyelin |
| CAS | 85187-10-6 |
| Color | White |
| Purity Grade Notes | Research |
| Quantity | 5 mg |
| Solubility Information | Ethanol : 25 mg/mL (34.15 mM; Need ultrasonic) |
| SMILES | [Sphingomyelin] |
| Molecular Weight (g/mol) | 732.08 |
| Formula Weight | 732.08 |
| Percent Purity | 95.0% |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction