Learn More
3M™ Chemical Spill Kit Dispenser
Ideal for rapid response to chemical spill containment and cleanup. 3M™ Chemical Spill Kit Dispenser contains 4 kits that include 2 Multi-Format sheets, Nitrile gloves, dustpan and brush, disposable bag and tie. Dispensers can be wall or shelf mounted for immediate access in areas where spills are likely to occur.
Brand: 3M™ 7100090373
162.93 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : 04
Additional Details : Weight : 1.58220kg
Descrizione
Ideal for rapid response to chemical spill containment and cleanup.- Cleaning up hazardous chemical spills, drips, and leaks
- Suitable for use with most chemicals
- Promotes safety on the site
- Easliy disposable
- Absorbs up to 1.5L per kit.
- Dispensers can be wall or shelf mounted for immediate access in areas where spills are likely to occur.
- 4 x disposable spill kits with dispenser
- 2 x Multi-Format sheets
- Nitrile gloves
- Dustpan and brush
- Disposable bag and tie
Specifica
1.5 L | |
Spill Kit Dispenser | |
4 disposable spill kits with dispenser, 2 Multi-Format sheets, one pair Nitrile gloves, Dustpan and brush, disposable bag and tie. | |
1 Ea. |