missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Spill Trough Laboratory Tray

Suited for environmentally dangerous aggressive substances, for laboratory work, as an etching tray, an instrument holder, a medicine tray, for photographic development, as a sample presentation tray or a display tray for foodstuffs. Buerkle™ Spill Trough Laboratory Trays are practical, all-purpose trays that can be nested when empty, and are durable, shatterproof and resist flexing.

Brand:  Buerkle™ 4201-1318

5.79 GBP valid until 2024-12-31
Use promo code "21745" to get your promotional price.


 View more versions of this product

Product Code. 10130501

  • £16.55 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

These laboratory trays are practical, all-purpose trays that can be nested when empty, and are durable, shatterproof and resist flexing. Also suitable as a spill trough for environmentally dangerous aggressive substances, for laboratory work, as an etching tray, as an instrument holder, a medicine tray, for photographic development, as a sample presentation tray or a display tray for foodstuffs.
  • Polypropylene, white
  • Manufactured using injection moulding, not by deep drawing
  • Trays can be nested
  • Tray not suitable for temperature controlled cupboards or dishwashers
  • Temperature resistant up to +60°C (briefly up to +80°C)
Specifications

Specifications

1 L
180 x 230 x 42 mm
42 mm (Outer Rim)
Polypropylene
180 mm (Inner Base), 190 mm (Outer Base), 230 mm (Outer Rim)
140 x 190 mm (base)
White
130 mm (Inner Base), 140 mm (Outer Base), 180 mm (Outer Rim)
Laboratory Tray
Rectangular
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers