missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ SpillGuard Leak Detection System
Shop All asecos Products
Click to view available options
:
SpillGuard Leak Detection System
Description
- Leak recognition system for ATEX zone 0
- Highly resistant and electrically conductive housing
- Acoustic and optical alarm for min. 24 hours in case of contact with hazardous liquid substances
- Regular optical signal (red LED) indicates safe function
- Acoustic signal indicates the need for replacement in good time at the end of the service life (up to 5 years)
- ATEX marking: EX II 1G Ex ia IIC T4 Ga
- External dimensions Dia.xH (mm): 110 x 40
Specifications
Specifications
| Type | Spill Containment System |
| Weight (English) | 0.32 lb. |
| Weight (Metric) | 0.145 kg |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion