missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Spin-X™ UF Concentrators
GREENER_CHOICE

Product Code. 11303795 Shop All Corning Products
Click to view available options
MWCO:
10 kDa
100 kDa
30 kDa
5 kDa
50 kDa
Quantity:
12/Pk.
25/Pk.
Volume (Metric):
500 μL
6 mL
20 mL
Filtration Area:
0.5 cm2
2.5 cm2
6 cm2
Unit Size:
Pack of 12
Pack of 25
This item is not returnable. View return policy

Product Code. 11303795

Brand: Corning™ 431479

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Disposable, single use only ultrafiltration devices with polyethersulfone membranes (PES) for the centrifugal concentration and/or purification of biological fluids and aqueous solutions

Spin-X UF centrifugal concentrators offer a simple, one step procedure for concentrating or desalting proteins and other biomolecules with 90% or better recovery.

  • The vertical membrane design and thin channel filtration chamber minimizes membrane fouling and provides fast, high speed concen­trat­ing, even with particle laden solutions.
  • Low binding polyethersulfone (PES) membranes are available with five molecular weight cut-offs (MWCO): 5,000, 10,000, 30,000, 50,000, and 100,000 to meet all of your concentrating needs. Choose an MWCO half to a third smaller than the protein to be concentrated.
  • The MWCO and graduations are printed on the side of the concentrator tube to avoid mix-ups.
  • Spin-X UF 6 and 20 concentrators can be used with either swinging bucket or fixed angle rotors. Spin-X UF 500 con­centrators require fixed angle rotors.
TRUSTED_SUSTAINABILITY

Specifications

Cap Material Polypropylene
Color White
Filter Type Centrifuge Filter
For Use With (Equipment) Fixed Angle Rotor
Material PES
Material (Membrane) PES
MWCO 30 kDa
Filtration Area 0.5 cm2
For Use With (Application) Concentration, desalting of proteins, enzymes, monoclonal antibodies, immunoglobulins, removal of labeled amino acids and nucleotides, HPLC sample preparation, deproteinization of samples, recovery of biomolecules from cell culture supernatants, lysates
Quantity 25/Pk.
Type Ultrafiltration Concentrator
Volume (English) Working 0.016 oz.
Volume (Metric) 500 μL
Volume (Metric) Working 0.5 mL
Height (English) 1.96 in.
Height (Metric) 5 cm
Diameter (English) 0.43 in.
Diameter (Metric) 1.1 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.