Learn More
Corning™ Spin-X™ UF Concentrators
GREENER_CHOICE
Disposable, single use only ultrafiltration devices with polyethersulfone membranes (PES) for the centrifugal concentration and/or purification of biological fluids and aqueous solutions
Brand:  Corning™ 431480
 View more versions of this product
Product Code. 11313795
  • £129.00 / Pack of 25
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Spin-X UF centrifugal concentrators offer a simple, one step procedure for concentrating or desalting proteins and other biomolecules with 90% or better recovery.
  • The vertical membrane design and thin channel filtration chamber minimizes membrane fouling and provides fast, high speed concen­trat­ing, even with particle laden solutions.
  • Low binding polyethersulfone (PES) membranes are available with five molecular weight cut-offs (MWCO): 5,000, 10,000, 30,000, 50,000, and 100,000 to meet all of your concentrating needs. Choose an MWCO half to a third smaller than the protein to be concentrated.
  • The MWCO and graduations are printed on the side of the concentrator tube to avoid mix-ups.
  • Spin-X UF 6 and 20 concentrators can be used with either swinging bucket or fixed angle rotors. Spin-X UF 500 con­centrators require fixed angle rotors.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polypropylene
Centrifuge Filter
PES
50 kDa
Concentration, desalting of proteins, enzymes, monoclonal antibodies, immunoglobulins, removal of labeled amino acids and nucleotides, HPLC sample preparation, deproteinization of samples, recovery of biomolecules from cell culture supernatants, lysates
Ultrafiltration Concentrator
500 μL
1.96 in.
0.43 in.
White
Fixed Angle Rotor
PES
0.5 cm2
25/Pk.
0.016 oz.
0.5 mL
5 cm
1.1 cm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Corning™ Spin-X™ UF Concentrators > 50,000 MWCO; 500 μL