missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ SPINeasy™ DNA Pro Kit for Soil
Isolation of microbial DNA from high and low biomass soil samples. Unbiased lysis of microbial populations for more accurate microbiome studies. Inhibitor removal technology extracts nucleic acids free of most notorious PCR inhibitors.
Brand: MP Biomedicals™ 116546050
This item is not returnable.
View return policy
Description
- The SPINeasy DNA Pro Kit for Soil has been carefully designed for the isolation of pure microbiome genomic DNA from challenging soil types, including those with low biomass or that are highly contaminated. Soil samples are complex environments characterized by the presence of inhibitory compounds, such as humic acid, heavy metals, and other aromatic components, which often prove to be challenging for downstream molecular analysis.
- The SPINeasy DNA Pro Kit for Soil effectively lyses microbial populations, including bacteria, fungi, viruses, and protists. The kit provides similar yields to that of our highly cited FastDNA™ SPIN Kit, but with improved purity and reduced processing time. Isolated DNA products showed no inhibition in PCR and were immediately ready to be used for downstream applications, including long fragment PCR, qPCR, and next-generation sequencing (16S and whole genome), without the need of further inhibitor removal steps.
Specifications
Store at Room Temperature, Dry Environment | |
Up to 500 mg of Soil Samples | |
50 Preps |
Spin Column | |
50x Lysing Matrix YB, 2 mL RNase A Solution, 55 mL Buffer S1, 22 mL Buffer S2, 50 mL Buffer S3, 28 mL Buffer S4, 40 mL Buffer S5, 6 mL Buffer S6, 50x Column S, 50x 2 mL collection tubes, 50x 1.5 mL collection tubes |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- SPINeasy DNA Pro Kit for Soil
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 C
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H336-May cause drowsiness or dizziness.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P321-Specific treatment (see on this label).
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : sodium thiocyanate,Isopropanol, Guanidine Thiocyanate, Ethanol
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.