Learn More
MP Biomedicals™ SPINeasy RNA Kit for Bacteria
SPINeasy RNA Kit for Bacteria - Quick and convenient purification of total RNA from gram-positive and gram-negative bacteria using spin column technology.
Brand:  MP Biomedicals™ 116541050
Additional Details : Weight : 0.20000kg
Product Code. 17782438
  • £414.46 / 50 sets
Estimated Shipment: 29-07-2024
to see stock.
Description
Description
  • SPINeasy RNA Kit for Bacteria is a silica-membrane spin-column kit that enables quick and convenient purification of total RNA from gram-positive and gram-negative bacteria. Included in the kit is our specially formulated RNASS solution that stabilizes and protects RNA in bacteria samples.
  • The use of Lysis Buffer R and Lysing Matrix B in combination with FastPrep™ instruments from MP Biomedicals enables highly efficient lysis of bacterial samples within seconds.
  • With a simple workflow, the kit allows multiple samples to be processed:
    • Simultaneously
    • Without the use of toxic substances such as phenol and chloroform.
  • Total RNA of high quality and integrity can be purified typically:
    • Within 40 minutes
    • Is immediately available for RT-PCR and other downstream applications.
Specifications
Specifications
Lysis Buffer
40 min.
Bacteria
Silica Spin Column
E. coli
50 Preps