Learn More
MP Biomedicals™ SPINeasy RNA Kit for Tissue
SPINeasy RNA Kit for Tissue - Quick and convenient purification of total RNA from animal tissues, plant tissues and tissue cultures using spin column technology.
Brand: MP Biomedicals™ 116543050
393.74 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Additional Details : Weight : 0.20000kg
Description
- SPINeasy RNA Kit for Tissue is a silica-membrane spin-column kit that enables quick and convenient purification of total RNA from:
- Various animal tissues
- Plant tissues
- Tissue cultures.
- The use of our specially formulated Lysis Buffer R and Lysing Matrix A in combination with FastPrep™ instruments from MP Biomedicals enables highly efficient lysis of tissue samples within seconds. With a simple workflow, the kit allows multiple samples to be processed:
- Simultaneously
- Without the use of toxic substances such as phenol and chloroform.
- Purified RNA is of high quality and integrity and immediately ready for RT-PCR and other downstream applications.
Specifications
Lysis Buffer | |
40 min. | |
Tissue |
Silica Spin Column | |
Human, Mouse, Rat | |
50 Preps |
Safety and Handling
- SPINeasy RNA Kit for Tissue
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H373-May cause damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P321-Specific treatment (see on this label).
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Manganese Sulfate Monohydrate, Calcium Chloride, Guanidine Hydrochloride