missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spironolactone, Tocris Bioscience™
Click to view available options
Quantity:
50 mg
Description
Competitive mineralocorticoid (aldosterone) receptor antagonist that exhibits antihypertensive activity in vivo. Also displays antiandrogen activity and inhibits steroid hormone biosynthesis.
Specifications
CAS | 52-01-7 |
Molecular Formula | C24H32O4S |
Quantity | 50 mg |
Synonym | spironolactone, aldactone, spirolactone, verospiron, euteberol, spiroctan, spirolang, verospirone, aldactone a, spironocompren |
InChI Key | LXMSZDCAJNLERA-ZHYRCANASA-N |
SMILES | CC(=O)SC1CC2=CC(=O)CCC2(C3C1C4CCC5(C4(CC3)C)CCC(=O)O5)C |
IUPAC Name | S-[(7R,8R,9S,10R,13S,14S,17R)-10,13-dimethyl-3,5'-dioxospiro[2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthrene-17,2'-oxolane]-7-yl] ethanethioate |
Molecular Weight (g/mol) | 416.576 |
PubChem CID | 5833 |
ChEBI | CHEBI:9241 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'