missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Split Stopper for Serum Bottles, Small

Labconco™ Split Stopper for Serum Bottles, Small are packs of 13mm rubber stoppers that fit in 2mL and 3mL glass serum bottles. 100/Pack

Brand:  Labconco™ 7576010

Code : 50

Additional Details : Weight : 0.45000kg

Product Code. 15183065

  • £38.60 / Pack of 100
Estimated Shipment: 06-09-2024
to see stock.
Add to basket

Description

Description

  • Serum Bottles hold small samples for batch drying on a tray
  • They may also be connected to valve ports on drying chambers and manifolds
  • Split stoppers sold separately for easy sealing
Specifications

Specifications

Split stopper
Drying small samples
Rubber
13 mm
Serum Bottle Stopper
100Pack
2 mL and 3 mL Serum Bottles
Flat Grip, Split Stem
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers