Learn More
Nabertherm™ Split Type Horizontal Tube Furnace RSH 1100 Series with B500 Controller
The split-type design makes it easy to change the working tube. It allows for a comfortable exchange of various working tubes.
£4422.91 - £10136.54
Product Code | Brand | Diameter (Metric) Outer | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outer | Length (Metric) | Price | Quantity & Availability | ||||
17388149
|
Nabertherm™
RS152L2AN |
50 mm | 250 mm |
£4422.91
Each |
Estimated Shipment: 17-09-2024
Log in to see stock. |
||||
17308159
|
Nabertherm™
RS252L2AN |
50 mm | 500 mm |
£4853.15
Each |
Estimated Shipment: 17-09-2024
Log in to see stock. |
||||
17348159
|
Nabertherm™
RS322L2AN |
120 mm | 500 mm |
£6204.11
Each |
Estimated Shipment: 17-09-2024
Log in to see stock. |
||||
17388159
|
Nabertherm™
RS382L2AN |
80 mm | 500 mm |
£5988.99
Each |
Estimated Shipment: 17-09-2024
Log in to see stock. |
||||
17328169
|
Nabertherm™
RS422L2AN |
120 mm | 750 mm |
£8045.56
Each |
Estimated Shipment: 17-09-2024
Log in to see stock. |
||||
17368169
|
Nabertherm™
RS472L2AN |
170 mm | 750 mm |
£9086.75
Each |
Estimated Shipment: 17-09-2024
Log in to see stock. |
||||
17308179
|
Nabertherm™
RS482L2AN |
80 mm | 750 mm |
£7658.34
Each |
Estimated Shipment: 17-09-2024
Log in to see stock. |
||||
17348179
|
Nabertherm™
RS522L2AN |
120 mm | 1000 mm |
£8708.13
Each |
Estimated Shipment: 17-09-2024
Log in to see stock. |
||||
17388179
|
Nabertherm™
RS572L2AN |
170 mm | 1000 mm |
£10136.54
Each |
Estimated Shipment: 17-09-2024
Log in to see stock. |
||||
Description
- Maximum Temperature: 1100°C
- Dual shell housing made of sheets of textured stainless steel
- Only fiber materials are used which are not classified as carcinogenic according to TRGS 905, class 1 or 2
- Type N thermocouple
- Split-type design for simple insertion of the working tube
- Working tube made of material C 530 including fiber plugs for operation under air in scope of delivery
- Heating elements on support tubes provide for free radiation
- Switchgear and control unit integrated in furnace housing
- Defined application within the constraints of the operating instructions
- NTLog Basic for Nabertherm controller: recording of process data with USB-flash drive
Specifications
B500 | |
Up to 1100°C |