missing translation for 'onlineSavingsMsg'
Learn More

Sponge Cobalt, A-8B46, promoted with Nickel and Chromium, Thermo Scientific Chemicals

£61.90 - £198.00

Chemical Identifiers

MDL Number MFCD00239945
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11346289
SDS View Documents
Thermo Scientific Alfa Aesar
044871.22
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11356289
SDS View Documents
Thermo Scientific Alfa Aesar
044871.36
500 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Sponge Cobalt, A-8B46, promoted with Nickel and Chromium, is used in synthetic chemistry for hydrogenation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

MFCD00239945

Specifications

MFCD00239945
Insoluble in water.
Slurry
Videos
Safety and Handling

Safety and Handling

P201-P202-P210-P223-P231+P232-P240-P241-P260-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P335+P334-P342+P311-P363-P370+P378q-P501c

H228-H261-H317-H334-H350-H373

missing translation for 'dotInformation' : Hazard Class: 4.2; Packaging Group: II

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sponge Cobalt, A-8B46, promoted with Nickel and Chromium, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.