Learn More
uvex Sport Occupational Black PU 65938 Low Shoe - Width 11
Product Code. 17212671 Shop All Uvex Products
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
49
50
51
52
Unit Size:
1 pair
Product Code. 17212671
Brand: uvex 6593850
Only null left
Add to basket
Add to basket
Offers first-class wearer comfort; low weight, climate-optimizes high-tech materials and ergonomic outsole
Uvex 1 Sport Shoe 6593.8 O1 FO SRC
  • Modern, ultra-lightweight and flexible O1 occupational shoe without toe cap
  • Made from synthetic materials, suitable for people allergic to chrome
  • All sole materials are free of silicones plasticizers and other paint wetting impairment substances
  • Meets ESD specification with leakage resistance less than 35 megohms
  • Ergonomically designed outsole comprising dual-density polyurethane with excellent slip resistance
  • Made with new developed technology for exceptional wearer comfort, climate-optimizes and breathable materials
  • Virtually seam-free construction made from high-tech materials to eliminate pressure points
  • Removable anti-static comfortable insole with moisture transport system and additional shock absorption around heel and forefoot
  • Soft, padding on collar and tongue
  • For light applications
  • Certified with Uvex medicare for individual, orthopedic inlay soles and individual adjustments of sole unit such as overall increase, inside and outside increase and other solutions
  • Article No. 6593.8
  • Standard EN ISO 20347:2012 O1 FO SRC
  • Sizes: 35 to 52
TRUSTED_SUSTAINABILITY
Specifications
Color Black
Material Textile
Size 50
Sole Material Dual-density polyurethane (PU/PU)
Toe Material Plain Toe
Disposable Reusable
Certifications/Compliance (Europe) EN ISO 20347:2012
Gender Unisex
Fit Standard Width
Model Uvex 1
Latex Free Yes
Waterproof No
EN ISO 20347 O1
Slip Resistance Rating SRC
Product Type ESD Safety Shoe