missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SPP 86
Potent RET inhibitor
£153.34 - £593.94
Specifications
Purity | >98% |
---|---|
Description | SPP 86 |
Molecular Weight (g/mol) | 277.32 |
Product Type | SPP 86 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18707088
|
Tocris Bioscience™
5391/50 |
50 mg |
£593.94
50mg |
Estimated Shipment: 29-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18770498
|
Tocris Bioscience™
5391/10 |
10 mg |
£153.34
10mg |
Estimated Shipment: 29-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Potent RET inhibitor (IC50 = 8 nM). Also exhibits inhibitory activity at EphA1, FGFR1, Flt4, Lck and Yes. Inhibits proliferation of HCT 116 and TPC1 cells in vitro. Cell permeable.Specifications
>98% | |
SPP 86 | |
277.32 | |
SPP 86 |
For Research Use Only