Learn More
GVS SPR580 Elipse Mask with ABE1P3
GREENER_CHOICE
Product Code. 18439675
Click to view available options
:
GVS SPR580 with ABE1P3
Product Code. 18439675
Brand: GVS SPR580
Only null left
Add to basket
Add to basket
This mask is recommended for multigas and dust risks (excluding ammonia), in chemical production and handling environment. Suitable for all types of organic gases, inorganic gases, formaldehyde, chemical gases, and dust.
This mask is recommended for multigas and dust risks (excluding ammonia), in chemical production and handling environment. Suitable for all types of organic gases, inorganic gases, formaldehyde, chemical gases, and dust.
  • Extremely lightweight and compact to give the wearer as much comfort as possible. It offers a full range of vision without interfering with other eye or ear protection which users are required to wear.
  • Low breathing resistance to reduce the risk of user fatigue and to add additional comfort.
  • Made from soft thermoplastic odorless elastomer that is hypoallergenic (without latex and silicone) and an easy-to-adjust headband.
  • Certifications: Mask Body EN140: 1998.
  • ABE1P3 R D Filters: EN14387: 2004 + A1: 2008. CE 2798
Specifications
Color Blue
Disposable Reusable