Learn More
Spreader Blue Polystyrene Wedge-Shaped Spreader
High impact S-shape disposable bacterial spreaders
Brand: Microspec D3
Code : 03
Additional Details : Weight : 0.70920kg
Description
High impact polystyrene disposable bacterial spreaders. Wedge shaped to reduce tendency to flip, or traditional “L” shape. Packs of 500 contain re-sealable inner bags of 20 sterile spreaders.Specifications
Yes | |
L-Shape | |
Blue | |
Polystyrene |
Sterile | |
Spreader | |
25 Bags of 20 | |
High impact Polystyrene disposable bacterial spreaders |