missing translation for 'onlineSavingsMsg'
Learn More
Learn More
De Buyer™ Spring Tongs
Grip, move and transfer food using the stainless steel De Buyer™ Spring Tongs
Brand: De Buyer™ 4788.24N
Code : A0
Additional Details : Weight : 0.09900kg
Description
- For general purpose food handling needs
- Clamp INOX
- Stainless Steel
- Three lengths
Specifications
Clamp Inox | |
Stainless Steel | |
240 mm | |
Each |