missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beckman Coulter™ SPRIselect™
Purify DNA fragments of the desired size for library preparation. Beckman Coulter™ SPRIselect™ chemistry speeds and simplifies nucleic acid size selection using paramagnetic bead-based chemistry for next-generation sequencing.
£406.50 - £1513.50
Specifications
Content And Storage | 15-30°C |
---|---|
Form | Liquid |
Format | Liquid |
For Use With (Equipment) | Automated processing systems, e.g. Biomek |
pH | 8.0 - 8.4 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15676104
|
Beckman Coulter™
B23317 |
5mL |
£406.50
5mL |
Estimated Shipment: 08-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15605838
|
Beckman Coulter™
B23318 |
60mL |
£1513.50
60mL |
Estimated Shipment: 08-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Proprietary SPRI paramagnetic bead-based chemistry provides:
- Tunable fragment size selection to suit specific applications
- Predictable, consistent size selection between runs and reagent lots
- Compatibility with manual and automated processing
- Customizable size selection: from 150 to 800 base pairs
- Suitable for manual or automated workflows; can be run on Biomek platforms
Specifications
15-30°C | |
Liquid | |
8.0 - 8.4 | |
Nucleic Acid |
Liquid | |
Automated processing systems, e.g. Biomek | |
Standard PCR |