missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Suitable for wearers of prescription glasses.
- Upper protection.
- Adjustable temples.
- Lower protection.
- EN 166: Guaranteed minimum resistance of the protective eyewear against everyday risks including dropping the protective eyewear onto the ground, ageing by light exposure, exposure to heat or corrosion etc.
- EN 170, Risks relating to radiation - UV protection: The protection filters ultraviolet radiation. This type of protection is particularly recommended during activities where the source of ultraviolet radiation is predominant, such as mercury vapor lamps, actinic or germicides. Levels of application: 2-1.2, 2C-1.2 Lens color: Clear, blue, yellow or green. Adding the code 'C' indicates good color recognition by the holder.
- EN 172, Solar radiation risks: Filter protection for industrial solar radiation. Recommended for outdoor work, EN172 norm guarantee a protection of 99.99% from ultra-violet solar radiation. Levels of application: 5-3.1 (Polarized) 5-2 (Twilight), 5-1.4 (ESP or CSP), 5/1.1-1.4 (Contrast), 5-2.5 and 5 to 4.1 (smoke) Lens color: depends on the lens technology.
- EN 175, Standard for welding: Specifies safety requirements and test methods related to the eye and face protective equipment used for welding and associated techniques. This European standard specifies the requirements and test methods related to the personal protective equipment used to protect the operator's eyes and face against harmful optical radiation and other specific risks due to customary welding or cutting processes or other associated techniques. The standard specifies protection, including ergonomic aspects, against the risks or hazards of various kinds: radiation, inflammability, mechanical and electrical risks. This standard defines the terms used and specifies the requirements related to materials, design and manufacture.
- Mechanical risk S - Extra strong, resists a 22 mm 43 g ball falling 1.30 m at 5,1 m/s.
- Optical class: Class 1, continuous work, worn permanently.
- CE, European standard: Frame marked to indicate meeting CE standard.

Specifications
Specifications
Certifications/Compliance | EN166, EN170, EN172/5, EN175, Mechanical risk S, Optical class 1, CE |
Lens Coating | Impact-resistant, Scratch-resistant, UV-treated |
Description | Model SQUWPCC5 |
Lens Tint | Shade 5 |
Product Type | Safety glasses |
For Use With (Application) | Welding, protection, continuous wear |
No. per Case | 180/Cs. |
No. per Pack | 1/bag, 10/Pk. |
Weight (Metric) | 47 g |
Lens Material | Polycarbonate |
Show More |
Documents
Product Information
Product Information
Bolle Safety SQUALE
File size: 626 kB
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Bollé Safety Squale Over the Glasses Safety Glasses >
Spot an opportunity for improvement?Share a Content Correction