missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Juchheim Laborgeraete™ Square Bosshead
Click to view available options
Height (Metric):
26 mm
Unit Size:
Each
Description
- Particularly versatile
- Very compact and aesthetic laboratory sleeve
- Max. Opening: 13 mm
- Made of particularly robust and durable malleable cast iron
- Coated with acid- and base-resistant epoxy resin powder varnish
- Made in Germany
Specifications
Specifications
| Material | Tempered Cast Iron |
| Height (Metric) | 26 mm |
| Weight (Metric) | 56 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction