Learn More
Gilac™ Square Bucket with Watertight Lid, 12 L
Square 12 L Bucket with Watertight Lid fit for food contact with watertight lid and pouring spout.
Brand: Gilac™ G615711
Additional Details : Weight : 1.20500kg
Description
- Bucket fit for food contact with watertight lid and pouring spout.
- Airtight lid keeps flavours in and odours out.
- Nesting when empty, no suction and stacking when lid is closed.
- Enables liquid preparations to be stored and transported.
- Graduation in litres.
- Discharge area that prevents water retention when being cleaned and anti-wear skirt that lifts the bucket off the ground.
- HDPE body and LDPE lid.
- Stainless steel handle.
- External dimensions include handle.
Specifications
Bucket, Handle, and Lid | |
12 L |